Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Total Dietary Fiber Reagant Kit

Catalog No. M1129790001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Total Dietary Fiber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
M1129790001 Total Dietary Fiber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1129790001 Supplier MilliporeSigma™ Supplier No. 1.12979.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Alpha-amylase solution (5mL), Protease solution (5mL), Amyloglucosidase solution (3 x 5mL)

For determination of Total Dietary Fiber in foods.

  • Enzymatic-gravimetric method
  • Packing: Fiber case
  • Sufficient for 100 tests
TRUSTED_SUSTAINABILITY

Specifications

Type Total Dietary Fiber
Includes alpha-amylase solution (5mL), Protease solution (5mL), Amyloglucosidase solution (3 x 5mL)
Packaging Fiber case
Storage Requirements 2°C to 8°C
Test Method Enzymatic-gravimetric method
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.