Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TRAIL, Human, Recombinant Protein, E. coli

Catalog No. 616374100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-637-4100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-637-4100UG Supplier MilliporeSigma™ Supplier No. 616374100UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, human TRAIL (amino acids 114-281) expressed in E. coli. Ligand for TRAIL receptors DR4 and DR5. Displays potent antitumor activity against selected targets and induces apoptosis in a number of transformed cell lines, but not in normal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 500mM NaCl, 10mM Na2HPO4, 2.7mM KCl, 2mM KH2PO4, 0.1mM DTT, ≤10% glycerol.
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Immunogen TRAIL
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.