Learn More
MilliporeSigma™ TRAIL (TNF-Related Apoptosis Inducing Ligand) Human Recombinant Protein
Shop All MilliporeSigma ProductsSpecifications
Specifications
| For Use With (Application) | Cell Culture |
| Name | Human Recombinant Protein, TRAIL (TNF-Related Apoptosis Inducing Ligand) |
| Quantity | 50 μg |
| Source | E. coli |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | Greater than 98% by SDS-PAGE/HPLC analyses |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.