Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 150mM NaCl, 20mM sodium phosphate buffer, pH 7.4. |
| Preparation Method | H2O (10mg/mL) |
| Purification Method | SDS-PAGE |
| Quantity | 100 mg |
| Immunogen | Transferrin, Holo |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or add NaN3 and refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 2 weeks at 4°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.