Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Trimethyl-Histone H3 (Lys9)α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710242MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-42MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-42MI Supplier MilliporeSigma™ Supplier No. 1710242
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Trimethyl-Histone H3 (Lys9)α Clone: in Human, Vertebrate samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Histone proteins are highly post-translationally modified however Histone H3 is the most extensively modified of the five histones. Histone H3 sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes. Trimethylation of histone H3 on Lys9 (H3K9me3) is one of the most highly studied epigenetic marks. H3K9me3 functions in the repression of euchromatic genes, and in epigenetic control of heterochromatin assembly, most likely by acting as a recognition motif for the binding of chromatin-associated proteins, such as Swi6 or HP1α. The enzymes responsible for H3K9 trimethylation are SUV39H1 and SUV39H2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trimethyl-Histone H3 (Lys9)α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Trimethyl-Histone H3 (Lys9) (mouse monoclonal). One vial containing 25μL of purified mouse monoclonal IgG1 in PBS with 0.05% sodium azide before the addition of glycerol to 30%. Normal Mouse IgG. One vial containing 25μg of purified IgG in 25μL of storage buffer containing 0.1% sodium azide. Control Primers, ZNF554. One vial containing 75μL of 5μM of each primer specific for the 3’ end of human ZNF554. FOR: CGG GGA AAA GCC CTA TAA AT; REV: TCC ACA TTC ACT GCA TTC GT
Gene Accession No. Q16695
Gene Symbols H3., H3/g, H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide corresponding to human Histone H3 trimethylated on Lys9.
Purification Method Protein G purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Test Specificity This antibody recognizes Histone H3 trimethylated at Lys9. Some cross-reactivity with dimethyl Lys9 is detected at higher concentrations, but the difference in specificity is >27-fold1. Phosphorylation of Ser10 interferes with antibody binding to trimethyl Lys91.
Includes This ChIPAb+ Trimethyl-Histone H3 (Lys9) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Vertebrates
Content And Storage 2°C to 8°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at -20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.