Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Trimethyl-Histone H3 (Lys9), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17625MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-625-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-625-MI Supplier MilliporeSigma™ Supplier No. 17625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Trimethyl-Histone H3 (Lys9) Clone: in Vertebrate samples, and it is validated for ChIP, Dot Blot, Western Blotting

The methylation of histones can occur on two different residues: arginine or lysine. Histone methylation can be associated with transcriptional activation or repression, depending on the methylated residue. Lysine 9 of histone H3 can be mono-, di- or trimethylated by different histone methyltransferases (HMTs) such as SuvH39H1 or G9a. This methylated lysine can be demethylated by histone demethylases as JMJD1A, LSD1 or JMJD2C. Methylation of this residue is mainly associated with transcriptional repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trimethyl-Histone H3 (Lys9)
Applications ChIP Assay, Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-trimethyl-Histone H3 (Lys9) (rabbit polyclonal IgG). One vial containing 100μg protein A purified IgG in 100μL of 0.02 M Phosphate buffer, pH 7.4, 0.25 M NaCl, 0.05% sodium azide with 30% glycerol. Normal Rabbit IgG. One vial containing 125μg of normal rabbit IgG in 125μL storage buffer containing 0.1% sodium azide. ChIP Primers, ZNF554. One vial containing 75μL of 5μM of each primer specific for ZNF554. FOR: CGG GGA AAA GCC CTA TAA AT; REV: TCC ACA TTC ACT GCA TTC GT
Gene Accession No. Q16695
Gene Symbols H3F3A, MGC87783, H3.3A, MGC87782, H3F3, H3.3B, H3F3B
Host Species Rabbit
Immunogen The trimethyl-histone H3 (Lys9) purified antibody is made against BSA-conjugated, synthetic peptide containing the sequence …AR[me3K]S… in which me3K corresponds to trimethyl lysine 9 of human Histone H3.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Test Specificity Recognizes histone H3, Mr 17kDa, trimethylated at lysine 9.
Includes Trimethyl-Histone H3 (Lys9) ChIP validated Antibody and Primer set including the ChIP-grade antibody and the specific control PCR primers used for chromatin immunoprecipitation of H3K9Me3.
Target Species Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 12 months from date of receipt; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.