Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Trypan Blue Dye for Microscopy

Catalog No. M1117320025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1117320025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1117320025 Supplier MilliporeSigma™ Supplier No. 1.11732.0025

for microscopy

Trypan blue is a dry dye for the preparation of a color-staining solution, staining histological sample material of human origin, e.g. trichrome staining in collagenous connective tissue.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
Storage Requirements 5° to 30°C
For Use With (Application) Histological staining
Melting Point >300°C (decomposition)
pH 9.8 at 10g/L, H2O, 20°C
Packaging Glass Bottle
CAS 72-57-1
Molecular Formula C34H24N6Na4O14S4
Formula Weight 960.79g/mol
Synonym Benzamine blue, Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt
Product Type Trypan Blue Dye
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.