Learn More
MilliporeSigma™ Tuerk's Solution for Leukocyte Counting
Catalog No. M1092770100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
M1092770100 100 mL
1 options
Catalog No. M1092770100 Supplier MilliporeSigma™ Supplier No. 1.09277.0100
Add to Cart
Add to Cart
for leukocyte counting
The counting of leukocytes is an important part of routine hematological diagnosis. The basis of all counting methods is the dilution and preparation of a blood sample of known volume. The erythrocytes are hemolyzed by the acetic acid of Tuerk’s solution and the leukocytes are stained by the dye contained. The required cell type in a defined volume is counted and the number of cells per microliter of blood is then calculated. Tuerk's solution is an IVD product and CE registered.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
For Use With (Application) Leukocyte Counting
Product Type Tuerk's Solution