Learn More
Specifications
Specifications
| For Use With (Application) | Histone Modification Assays, Chromatin Assembly Studies |
| Gene ID (Entrez) | NM_005321.2 |
| Name | Histone H1 protein |
| Purification Method | Sterilized through a 0.2 micron membrane filter |
| Quantity | 20 mg |
| Source | Lysine rich fraction from calf thymus |
| Storage Requirements | -20°C for 12 months. |
| Cross Reactivity | Bovine |
| Recombinant | Native |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.