Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Bovine Pituitary Extract

Catalog No. 02103MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
150 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-103MI 50 mg
02-104MI 150 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-103MI Supplier MilliporeSigma™ Supplier No. 02103
Add to Cart
Add to Cart

Designed for use as a cell culture media supplement

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stable for 2 years at 4°C from date of shipment. Rehydrated: Stable for 3 months at -20°C
For Use With (Application) Cell Culture
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.