Learn More
MilliporeSigma™ Upstate ChIPAb+ Dimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set
Catalog No. 17681MI Shop All MilliporeSigma Products
Change view
Click to view available options
Product Type:
ChIPAb+ Dimethyl-Histone H3 (Lys9) -ChIP Validated Antibody and Primer Set
Catalog No. Product Type
17-681-MI ChIPAb+ Dimethyl-Histone H3 (Lys9) -ChIP Validated Antibody and Primer Set
1 options
Catalog No. 17-681-MI Supplier MilliporeSigma™ Supplier No. 17681
Add to Cart
Add to Cart
Includes
Negative control mouse IgG antibody and control primers specific for human beta-globin promoter
Dimethyl-histone H3 (Lys9) purified antibody is made against synthetic peptide (dimethylated at Lys9) corresponding to amino acids 1-18 of histone H3
TRUSTED_SUSTAINABILITY
Specifications
Product Type ChIPAb+ Dimethyl-Histone H3 (Lys9) -ChIP Validated Antibody and Primer Set