Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Endothelial Cell Growth Supplements
GSA_VA

Catalog No. 02102MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
150 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-102MI 150 mg
02-101MI 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-102MI Supplier MilliporeSigma™ Supplier No. 02102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use as a cell culture media supplement

ECGS is mitogenic for many types of cells under reduced or serum-free conditions, such as mammalian, avian and human endothelial cells, smooth muscle cells, keratinocytes, melanocytes and hybridomas; ECGS often fully substitutes for feeder layers in culture of fastidious cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 years at 4°C; rehydrated: 3 months at -20°C
For Use With (Application) Cell Culture
Quantity 150 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.