Learn More
Specifications
Specifications
| Characterization | >98% purity by SDS PAGE |
| Concentration | 0.2μg/μL |
| For Use With (Application) | Chromatin Assembly Studies |
| Formulation | 20μg of mononucleosomes in buffer containing 20mM Tris-Cl pH 7.9, 1mM EDTA, 0.5mM PMSF,1mM DTT, and 50% glycerol. |
| Name | Native Nucleosomes |
| Purification Method | Glycerol gradient and ion exchange chromatography |
| Quantity | 20 μg |
| Source | HeLa cell nuclei |
| Storage Requirements | -80°C for 12 months |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.