Learn More
Specifications
Specifications
| For Use With (Application) | Western Blot, Enzyme Assay |
| Gene ID (Entrez) | NM_003538.3 |
| Name | Histone H4 Protein |
| Purification Method | FPLC agarose column |
| Quantity | 1 mg |
| Source | E. coli |
| Storage Requirements | Two years at -20°C from date of shipment. |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.