Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Rac/cdc42 Assay Reagent (PAK1 PBD, Agarose)

Catalog No. 14325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
300 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-325 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-325 Supplier MilliporeSigma™ Supplier No. 14325
Add to Cart
Add to Cart

Specifications

For Use With (Application) Affinity Binding Assay
Formulation Bound to glutathione agarose
Gene ID (Entrez) NM_044472.2, NM_001039802.1, NM_001791.3
Name PAK1 PBD fusion GST-tagged fusion protein
Quantity 300 μg
Source E. coli
Storage Requirements -20°C for 12 months.
Conjugate GST-Tag
Cross Reactivity Human
Recombinant Native
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.