Learn More
Specifications
Specifications
| For Use With (Application) | Affinity Binding Assay |
| Formulation | 650μg of recombinant protein in 1mL glutathione-agarose slurry containing 45mM TRIS, pH 7.5, 0.45% Triton X-100, 135mM NaCl, 4.5mM MgCl2, 0.9mM dithiothreitol, 0.09mM PMSF, 0.9μg/mL aprotinin, 0.9μg/mL leupeptin, and 10% glycerol. |
| Gene ID (Entrez) | NM_033046.2, NM_001015055.1, NM_001015056.1 |
| Name | Rhotekin RBD GST-tagged fusion protein |
| Quantity | 650 μg |
| Source | E. coli |
| Storage Requirements | -70°C for 6 months |
| Conjugate | GST-Tag |
| Cross Reactivity | Mouse |
| Recombinant | Native |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.