Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RIPAb+™ hnRNP C1/C2 RIP Validated Antibody and Primer Set

Catalog No. 03205MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-205MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-205MI Supplier MilliporeSigma™ Supplier No. 03205

missing translation for 'validMainframeMsg'

This RIPAb+ hnRNP C1/C2 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control normal mouse IgG antibody and control primers specific for the cDNA of human c-myc 3′ UTR.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Gene Accession No. NP_001070910.1
Gene Symbols hnRNPC, HNRNPC, C1, C2, HNRNP, HNRPC, SNRPC
Host Species Mouse
Immunogen RNP's eluted from oligo (dT) cellulose column of human HnRNP C1/C2.
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human, Mouse
Content And Storage -20°C for 12 months.
Product Type RIPAb+ hnRNP C1/C2 RIP Validated Antibody and Primer Set
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.