Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RIPAb+™ Lin28 RIP Validated Antibody and Primer Set

Catalog No. 03105MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-105MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-105MI Supplier MilliporeSigma™ Supplier No. 03105

This RIPAb+ Lin28 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Included negative control rabbit IgG antibody and control primers specific for CDK4.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoprecipitation
Classification Polyclonal
Description Negative control rabbit IgG antibody and control primers specific for CDK4
Gene Accession No. NP_078950
Gene Symbols ZCCHC1, Lin28, Lin28A, CSDD1
Host Species Rabbit
Immunogen Full-length His-tagged human Lin28 recombinant protein
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human, Mouse
Content And Storage -20°C for 12 months.
Product Type RIPAb+ Lin28 RIP Validated Antibody and Primer Set
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.