Learn More
MilliporeSigma™ Upstate™ TCF Reporter Plasmid Kit
Catalog No. 17285 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
17-285 1 Kit
1 options
Catalog No. 17-285 Supplier MilliporeSigma™ Supplier No. 17285
Add to Cart
Add to Cart
Set of transfection grade T cell factor (TCF) reporter plasmids for use in TOPFlash and FOPFlash wnt/b-catenin activity assays.
This gene encodes transcription factor 4, a basic helix-turn-helix transcription factor. The encoded protein recognizes an Ephrussi-box ('E-box') binding site ('CANNTG') - a motif first identified in immunoglobulin enhancers. This gene is expressed predominantly in pre-B-cells, although it is found in other tissues as well. Multiple alternatively spliced transcript variants that encode different proteins have been described.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit