Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
Shop All MilliporeSigma ProductsDescription
Control bacteria and oxidize organics
- Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
Specifications
| Product Type | UV Lamp |
| For Use With (Equipment) | Milli-Q Advantage a10 TOC Monitor |
| Wavelength | 254 nm |