Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Vanilloid Receptor Like Protein-1 (744-761), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. PC4211SET Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC4211SET 1 Set Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC4211SET Supplier MilliporeSigma™ Supplier No. PC4211SET

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Vanilloid Receptor Like Protein-1 (744-761) Clone: in Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immuno Blotting

The detection of noxious stimuli (chemical, mechanical, or thermal), a process referred to as nociception, occurs predominantly at the peripheral terminals of primary afferent neurons. This information is ultimately transmitted to the central nervous system to evoke a perception of pain which initiates appropriate protective reflexes. Vanilloid-containing compounds which include capsaicin, the active ingredient in hot chilli peppers, selectively depolarize nociceptors to activate the nociceptive pathway. The receptor for capsaicin, vanilloid receptor 1 (VR1), has been identified as a nonselective cation channel that resembles members of the transient receptor potential (TRP) family of ion channels. The vanilloid receptor 1 protein functions both as a receptor for capsaicin and a transducter of noxious thermal stimuli. Immunocytochemical and mRNA localization studies suggest that VR1 protein is localized to small-diameter sensory neurons within the dorsal root ganglia and nerve terminals in the dorsal horn. A capsaicin-receptor homolog has recently been identified, designated the vanilloid-receptor-like protein 1 (VRL-1). VRL-1 is activated by high temperatures and is expressed in medium- to large-diameter neurons within sensory ganglia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vanilloid Receptor Like Protein-1 (744-761)
Applications Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation 100μg antibody in 0.05 M sodium phosphate buffer, 0.2% gelatin and 25μg lyophilized control peptide (immunogen) (100μg size only).
Host Species Rabbit
Immunogen a synthetic peptide (KNSASEEDHLPLQVLQSP) corresponding to amino acids 744-761 of rat vanilloid receptor like protein 1, conjugated to KLH
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽98kDa (doublet) vanilloid receptor like protein-1 in rat spinal cord extract. Supplied with a control peptide (100μg size only).
Includes 100μg antibody in 0.05M sodium phosphate buffer, 0.2% gelatin and 25μg lyophilized control peptide (immunogen) (100μg size only).
Target Species Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.