Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Vasoactive Intestinal Peptide, Human, Porcine, and Rat, Calbiochem

Catalog No. 0523210105M Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-232-10105M 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-232-10105M Supplier MilliporeSigma™ Supplier No. 052321010.5MG
Acts as a mitogenic factor for embryonic neurons in the sympathetic nervous system. Has powerful hypotensive and vasodilatory effects. Behaves either as a circulating hormone or as a neurotransmitter. Stimulates cAMP production in rat peritoneal macrophages. Inhibits the growth of human small-cell lung carcinoma (SCLC) both in vitro and in vivo, suggesting that it may be useful as a less toxic alternative to conventional chemotherapeutic agents in the treatment of SCLC. Nitric oxide stimulates the release of this peptide.
TRUSTED_SUSTAINABILITY

Specifications

Species Human, Porcine, Rat
Purification Method HPLC
Purity ≥95%
CAS 40077-57-4
Color White to Off-white
Content And Storage Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Format Lyophilized Solid
Formulation Supplied as a trifluoroacetate salt.
Product Type Vasoactive Intestinal Peptide
Quantity 0.5 mg
Regulatory Status RUO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.