Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VEGFR1, Human, Recombinant Protein, S. frugiperda

Catalog No. PF08250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF08250UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF08250UG Supplier MilliporeSigma™ Supplier No. PF08250UG
Only null left
Add to Cart
Add to Cart
Recombinant, human VEGFR-1 expressed in S. frugiperda insect cells. Plays an important role in angiogenesis. Upon binding to VEGF, the receptor undergoes dimerization and is activated, leading to downstream signaling.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered PBS, 300mM NaCl, 50μg BSA/μg of cytokine.
Preparation Method Reconstitute to ≥10μg/mL with sterile PBS containing ≥0.1% HSA or BSA. It is recommended that the protein be titrated for optimal results in individual systems.
Purification Method SDS-PAGE
Quantity 50 μg
Source S. frugiperda
Immunogen VEGFR1
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.