Learn More
MilliporeSigma™ Wall Mounting Bracket for Washer Distribution Kit
Catalog No. WMBWASH1 Shop All MilliporeSigma Products
Change view
Click to view available options
Wall mounting bracket
Catalog No.
WMBWASH1 Wall mounting bracket
1 options
Catalog No. WMBWASH1 Supplier MilliporeSigma™ Supplier No. WMBWASH1
Add to Cart
Add to Cart
Reversible R/L mounting for use with PE reservoir
  • For mounting Washer Distribution Kit to a wall, adjacent to 30/60/100L pure water storage reservoir
  • For use with Elix™ Advantage and RiOs™ 3
TRUSTED_SUSTAINABILITY
Specifications
Type Wall Mounting Bracket
For Use With (Equipment) Elix™ Advantage, RiOs™ 3