Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Water/TFA 0.1% Mobile Phase

Catalog No. MTX1280P1 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX1280P1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX1280P1 Supplier MilliporeSigma™ Supplier No. TX1280P1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Filtered through 0.2μm filter

  • Clear liquid form
  • Assay (trifluoroacetic acid): 0.095 to 0.105% (v/v)
  • Molar mass is 114.02

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Quantity 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.