Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Wee1, GST-Fusion, Human, Recombinant Protein, S. frugiperda

Catalog No. 32589610UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-589-610UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-589-610UG Supplier MilliporeSigma™ Supplier No. 32589610UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, human Wee1 fused at the N-terminus to a GST-His6-thrombin cleavage site sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. Wee1 is a tyrosine kinase that causes inhibitory phosporylation of Cdk1 and Cdk2 and acts as a mitotic inhibitor. Protects the nucleus from cytoplasmically activated cyclin B1-complexed CDC2 prior to the onset of mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 100mM NaCl, 50mM Tris-HCl, 5mM DTT, 4mM reduced glutathione, 20% glycerol, pH 8.0.
Quantity 10 μg
Source S. frugiperda
Immunogen Wee1
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate GST-Fusion
Species Human
Recombinant Recombinant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.