Learn More
MilliporeSigma™ EMD Milllipore™ Elastica van Gieson Staining Kit
Catalog No. M1159740002 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
M1159740002 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. M1159740002 Supplier MilliporeSigma™ Supplier No. 1159740002
This item is not returnable. View return policy
Stains elastic fibers in tissue (Histological) sections
  • Connective tissue staining
  • Used to stain elastic fibers in tissue (Histological) sections
  • Used in conjunction with resorcinol-fuchsin solution according to Weigert
  • Results in visualization of black-brown nuclei, black-stained elastic fibers, red collagen yellow-stained muscles
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Storage Requirements 15° to 25°C