Learn More
TARGETMOL CHEMICALS INC EMD57033 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T25372
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. EMD57033 is a cardiac troponin C (cTnC) activator a dominant Ca2+ sensitizer which functions by binding to cardiac/slow skeletal troponin C heterodimers to promote cardiac contraction. purity: 99%
Catalog No. 50-258-0756