Learn More
Medchemexpress LLC HY-15193A 5mg Medchemexpress, EMD638683 (R-Form) CAS:1184940-47-3 Purity:>98%
SDP
Catalog No. 502081690
Encompass_Preferred
Medchemexpress, HY-15193A 5mg EMD638683 (R-Form) CAS:1184940-47-3 EMD638683 R-Form is the R-form of EMD638683. EMD638683 is a highly selective SGK1 inhibitor with IC50 of 3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1690 Supplier Medchemexpress LLC Supplier No. HY15193A5MG
Medchemexpress, HY-15193A 5mg EMD638683 (R-Form) CAS:1184940-47-3 EMD638683 R-Form is the R-form of EMD638683. EMD638683 is a highly selective SGK1 inhibitor with IC50 of 3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.