Learn More
Selleck Chemical LLC Emedastine Difumarate
SDP
Catalog No. 5000003046
Encompass_Preferred
Emedastine Difumarate (Rapimine Emedastine fumarate) is the difumarate salt form of emedastine a second generation selective histamine H1 receptor antagonist with anti-allergic activity
Catalog No. 50-000-03046 Supplier Selleck Chemical LLC Supplier No. S56475MG
Emedastine Difumarate (Rapimine Emedastine fumarate) is the difumarate salt form of emedastine a second generation selective histamine H1 receptor antagonist with anti-allergic activity