Learn More
Quality Biological Inc EMEM (1X) without Phenol Red, 500 mL
SDP
Catalog No. 507517645
Encompass_Preferred
EMEM (Eagle's Minimal essential Medium) is a cell culture medium developed by Henry Eaglt that can be used to maintain cells in tissue culture. Basal medium with different variations.
Without L-Glutamine
Without Phenol Red
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.
Catalog No. 50-751-7645 Supplier Quality Biological Inc Supplier No. 112212101
EMEM (Eagle's Minimal essential Medium) is a cell culture medium developed by Henry Eaglt that can be used to maintain cells in tissue culture. Basal medium with different variations.
Without L-Glutamine
Without Phenol Red
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.