Learn More
Med Vet International Emeraid intensIVe Care Hdn Canine, Highly Digestible Nutrition, 100gm
Supplier:  Med Vet International RXINTENSIVEC100GM
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Emeraid Intensive Care HDN Canine is highly digestible nutrition scientifically formulated for use in dogs with anorexia, critical illness, post-op in debilitated patients, anemia, and more. Its ingredients have been broken down into small, easy-to-absorb components. This elemental nutrition provides nutrients in small sizes like amino acids for easier and quicker assimilation of nutrients. The powdered formula is mixed with water to feed. High energy (2.26 to 1.36 kcal/ml ME) 1-month shelf life after opening Usable in tubes as small as 5 Fr without clogging All ingredients are optimized for assisting stressed patients, including amino acids, easily digestible protein, omega-3 fatty acids, purified fiber, cellulose, antioxidants, stabilized vitamins, chelated minerals, balanced electrolytes, and more Hydrolyzed starch supplies glucose, disaccharides, and oligosaccharides
Catalog No. 50-282-7908