Learn More
Med Vet International Emeraid Intensive Care Hdn Feline, Highly Digestible Nutrition, 100gm
Catalog No. 502828507
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Emeraid Intensive Care HDN Feline is highly digestible nutrition scientifically formulated for use in cats with anorexia, critical illness, post-op in debilitated patients, anemia, and more. Its ingredients have been broken down into small, easy-to-absorb components. This elemental nutrition provides nutrients in small sizes like amino acids for easier and quicker assimilation of nutrients. The powdered formula is mixed with water to feed.
Catalog No. 50-282-8507 Supplier Med Vet International Supplier No. RXINTENSIVEF100GM
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Emeraid Intensive Care HDN Feline is highly digestible nutrition scientifically formulated for use in cats with anorexia, critical illness, post-op in debilitated patients, anemia, and more. Its ingredients have been broken down into small, easy-to-absorb components. This elemental nutrition provides nutrients in small sizes like amino acids for easier and quicker assimilation of nutrients. The powdered formula is mixed with water to feed.