Learn More
Takara Bio EmeraldAmp® GT PCR Master Mix, 160 Rxns
SDP
Catalog No. 50444008
Encompass
This 2X PCR master mix is ideal for high-throughput screening. This convenient premix includes a green gel-loading dye and a density reagent. Reactions performed with this master mix can be loaded directly onto an agarose gel for electrophoresis. 160 Rxns
Catalog No. 50-444-008 Supplier Takara Bio Supplier No. RR310A
This 2X PCR master mix is ideal for high-throughput screening. This convenient premix includes a green gel-loading dye and a density reagent. Reactions performed with this master mix can be loaded directly onto an agarose gel for electrophoresis. 160 Rxns