Learn More
Takara Bio EMERALDAMP GT 800 RXN
SDP
Catalog No. 50444009
Encompass
PCR Master Mix, EmeraldAmp; 800 x 50uL rxns; Includes optimized buffer, PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix
Catalog No. 50-444-009 Supplier Takara Bio Supplier No. RR310B
PCR Master Mix, EmeraldAmp; 800 x 50uL rxns; Includes optimized buffer, PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix