Learn More
Takara Bio EMERALDAMP MAX 800 RXN
SDP
Catalog No. 50444012
Encompass
PCR Master Mix, EmeraldAmp Max; 800 x 50uL rxns; Includes optimized buffer, high-yield PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix; High yield: 10X more end product
Catalog No. 50-444-012 Supplier Takara Bio Supplier No. RR320B
PCR Master Mix, EmeraldAmp Max; 800 x 50uL rxns; Includes optimized buffer, high-yield PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix; High yield: 10X more end product