Learn More
Takara Bio EMERALDAMP MAX HS 160 RXN
SDP
Catalog No. 50444014
Encompass
PCR Master Mix, EmeraldAmp Max HS; 160 x 50uL rxns; Includes optimized buffer, high-yield hot start PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix; High yield: 10X more end product
Catalog No. 50-444-014 Supplier Takara Bio Supplier No. RR330A
PCR Master Mix, EmeraldAmp Max HS; 160 x 50uL rxns; Includes optimized buffer, high-yield hot start PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix; High yield: 10X more end product