Learn More
Takara Bio EMERALDAMP MAX HS 800 RXN
SDP
Catalog No. 50444015
Encompass
PCR Master Mix, EmeraldAmp Max HS; 800 x 50uL rxns; Includes optimized buffer, high-yield hot start PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix; High yield: 10X more end product
Catalog No. 50-444-015 Supplier Takara Bio Supplier No. RR330B
PCR Master Mix, EmeraldAmp Max HS; 800 x 50uL rxns; Includes optimized buffer, high-yield hot start PCR enzyme, dNTP mixture, gel loading dye (green) and density reagent in 2X premix; High yield: 10X more end product