Learn More
Pyramex™ Emerge™ Plus Safety Eyewear
Catalog No. 19155941 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Product Type:
Full Reader
Top Insert Reader
Magnification Power:
+1.50
+2.00
+2.50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Magnification Power
19-155-941 Full Reader +2.50
19-155-939 Full Reader +1.50
19-155-940 Full Reader +2.00
19-155-943 Top Insert Reader +1.50
19-155-944 Top Insert Reader +2.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-155-941 Supplier Pyramex™ Supplier No. SG9810R25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Emerge™ Plus Safety Eyewear features a clear polycarbonate lens with nylon frame.
  • Rubber temple tips
  • Soft nosepiece provides comfort for extended use
  • Translucent grey frame
  • Available in full readers or top insert readers
Specifications
Certifications/Compliance ANSI Z87.1, CSA Z94.3-07
Lens Coating Scratch-resistant, UV-treated
Lens Tint Clear
Product Type Full Reader
Frame Color Translucent Gray
Magnification Power +2.50
Weight (Metric) 31.3 g
Disposable Reusable
Lens Material Polycarbonate
Frame Material Nylon
UV Protection U6
Frame Type Full Frame