Learn More
Pyramex™ Emerge™ Safety Eyewear
Catalog No. 19155824 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Translucent Gray
Lens Tint:
Clear
IR/PC 3.0
IR/PC 5.0
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-824 Black Clear
19-155-825 Black IR/PC 5.0
19-155-826 Black IR/PC 3.0
19-155-938 Translucent Gray Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-155-824 Supplier Pyramex™ Supplier No. SB7910DRX
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Suitable for both working environments and sports activities
  • Stylish dual lens with full eye protection, plano lens, prescription lens, and full reader lens adaptable
  • Soft nose piece provides comfort for extended use
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Available with a full reader +1.5 or +2.0 lens (Emerge™ Prescription Safety Eyewear) or as a prescription* lens
    *NOTE: Emerge frame must be presented to your optometrist in order to be fitted with prescription lenses
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
    Description Will accommodate an Rx lens to the maximum of +3 to -3 diopters
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Black
    Magnification Power None
    Weight (Metric) 30 g
    Lens Material Polycarbonate
    Frame Material G300 Nylon