Learn More
MSC Vikan Collapsible Emergency Blade Shovel, ABS, 36.5 in, Black
Catalog No. 502837713
Encompass
This compact shovel is ideal for spill kit applications and various other uses. Constructed from injection-molded ABS plastic, it prevents damage to tires or truck beds. It is non-sparking, chemical, and corrosion resistant, making it a valuable addition to spill kits. The shovel collapses for easy storage in commercial trucks, vehicles, or even a family car.
  • Designed for commercial and industrial applications.
  • Collapsible handle for easy storage in commercial trucks, personal vehicles, and spill kits.
  • Made of high-quality injection-molded ABS plastic that will not damage tires or truck beds.
  • Handle and grip shapes are designed to minimize hand strain and improve tool control.
  • Suitable for both right- and left-handed users.
  • Non-sparking, chemical, and corrosion-resistant, making it ideal for use in spill kits.
Catalog No. 50-283-7713 Supplier MSC Supplier No. 41861766
This compact shovel is ideal for spill kit applications and various other uses. Constructed from injection-molded ABS plastic, it prevents damage to tires or truck beds. It is non-sparking, chemical, and corrosion resistant, making it a valuable addition to spill kits. The shovel collapses for easy storage in commercial trucks, vehicles, or even a family car.
  • Designed for commercial and industrial applications.
  • Collapsible handle for easy storage in commercial trucks, personal vehicles, and spill kits.
  • Made of high-quality injection-molded ABS plastic that will not damage tires or truck beds.
  • Handle and grip shapes are designed to minimize hand strain and improve tool control.
  • Suitable for both right- and left-handed users.
  • Non-sparking, chemical, and corrosion-resistant, making it ideal for use in spill kits.