Learn More
Grainger Haws Wall Mount Drench Hose, 8 ft. Hose, ABS Plastic, 1 ea.
Catalog No. 502791829
Encompass_Preferred
This single-head wall-mounted drench hose provides supplemental emergency body spray capabilities for environments requiring extended coverage. The unit features an 8 ft. (243.8 cm) reinforced rubber hose and a durable spray head with an integral flip-top dust cover to ensure operational readiness. Designed for efficiency, the system includes a stay-open squeeze lever valve for full continuous flow during emergency use.
  • Flow rate of 6.25 GPM (23.7 L/min)
  • Pressure rated to 250 psi (17.2 bar)
  • 1/2 in. NPT inlet connection
  • Chrome-plated brass stay-open squeeze lever valve with integral flow control
  • Certified to ANSI Z358.1 standards
  • Includes wall mounting brackets and a universal emergency sign
  • Requires a vacuum breaker for state code compliance
Catalog No. 50-279-1829 Supplier Grainger Supplier No. 16D414
This single-head wall-mounted drench hose provides supplemental emergency body spray capabilities for environments requiring extended coverage. The unit features an 8 ft. (243.8 cm) reinforced rubber hose and a durable spray head with an integral flip-top dust cover to ensure operational readiness. Designed for efficiency, the system includes a stay-open squeeze lever valve for full continuous flow during emergency use.
  • Flow rate of 6.25 GPM (23.7 L/min)
  • Pressure rated to 250 psi (17.2 bar)
  • 1/2 in. NPT inlet connection
  • Chrome-plated brass stay-open squeeze lever valve with integral flow control
  • Certified to ANSI Z358.1 standards
  • Includes wall mounting brackets and a universal emergency sign
  • Requires a vacuum breaker for state code compliance