Learn More
Grainger HAWS Emergency Mixing Valve, Brass, 12 GPM, 1/2 in. x 3/4 in. NPT
Catalog No. 502791832
Encompass_Preferred
This emergency valve thermostatically mixes hot and cold water to provide a safe, tempered fluid supply for emergency eyewash equipment. Featuring a low-lead brass construction, this unit ensures reliable performance through a fail-safe design that includes a secondary high-temperature limit valve for added protection against scalding. In the event of a hot water pressure loss or main thermostat malfunction, integrated cold water by-pass valves activate to maintain a continuous water supply.
  • Anti-scald protection
  • Flow range of 1 to 12 GPM provides service for one or two safety eyewashes
  • Shuttle design eliminates valve binding
  • Paraffin-filled thermostats offer industry-leading response times
  • Cold water by-pass capacity of 10 GPM
  • Certified to ANSI Z358.1, ASSE 1071, and NSF/ANSI 372 standards
  • Oversized valve seats resist failures caused by foreign matter
Catalog No. 50-279-1832 Supplier Grainger Supplier No. 16D420
This emergency valve thermostatically mixes hot and cold water to provide a safe, tempered fluid supply for emergency eyewash equipment. Featuring a low-lead brass construction, this unit ensures reliable performance through a fail-safe design that includes a secondary high-temperature limit valve for added protection against scalding. In the event of a hot water pressure loss or main thermostat malfunction, integrated cold water by-pass valves activate to maintain a continuous water supply.
  • Anti-scald protection
  • Flow range of 1 to 12 GPM provides service for one or two safety eyewashes
  • Shuttle design eliminates valve binding
  • Paraffin-filled thermostats offer industry-leading response times
  • Cold water by-pass capacity of 10 GPM
  • Certified to ANSI Z358.1, ASSE 1071, and NSF/ANSI 372 standards
  • Oversized valve seats resist failures caused by foreign matter