Learn More
Grainger Haws Emergency Shower, Fully Recessed, Ceiling Mount, 20 GPM, 1 ea
Catalog No. 502792229
Encompass_Preferred
This fully recessed, ceiling-mount emergency shower is designed for areas where individuals may be exposed to injurious or corrosive materials. It provides a reliable, plumbed solution for workplace safety, meeting ANSI/ISEA Z358.1 standards.
  • Features a hydrodynamic polished stainless steel showerhead with 20 gpm flow control for uniform water distribution.
  • Equipped with a brass stay-open ball valve with a stainless steel ball and stem to resist corrosion.
  • Includes a 111.8 cm (44 in.) stainless steel triangular pull rod for easy activation.
  • Constructed with 2.5 cm (1 in.) Schedule 40 hot-dipped galvanized steel piping for durability.
  • Supplied with a universal emergency sign and inspection tag for compliance and maintenance.
Catalog No. 50-279-2229 Supplier Grainger Supplier No. 16D381
This fully recessed, ceiling-mount emergency shower is designed for areas where individuals may be exposed to injurious or corrosive materials. It provides a reliable, plumbed solution for workplace safety, meeting ANSI/ISEA Z358.1 standards.
  • Features a hydrodynamic polished stainless steel showerhead with 20 gpm flow control for uniform water distribution.
  • Equipped with a brass stay-open ball valve with a stainless steel ball and stem to resist corrosion.
  • Includes a 111.8 cm (44 in.) stainless steel triangular pull rod for easy activation.
  • Constructed with 2.5 cm (1 in.) Schedule 40 hot-dipped galvanized steel piping for durability.
  • Supplied with a universal emergency sign and inspection tag for compliance and maintenance.