Learn More
Grainger HAWS Emergency Drench Shower, Freeze-Resistant, Wall-Mounted, Galvanized Steel, 1 ea
Catalog No. 502791818
Encompass_Preferred
This wall-mount, freeze-resistant emergency shower is designed for climates where temperatures can drop below freezing. The unit features supply lines and a pre-built brass valve assembly that install in a heated space on the opposite side of a wall to prevent freezing, and the system drains completely after use to avoid standing water. It utilizes a hydrodynamic showerhead design that delivers a more effective and comfortable flow by distributing water equally across the entire footprint.
  • Flow rate of 20 GPM
  • Includes universal emergency sign and inspection tag
  • Galvanized steel pipe with 1 in. NPT female inlet
  • 11 in. diameter ABS plastic showerhead
  • Reduced installation time with pre-built valve assembly
  • Certified to ANSI Z358.1 and CSA standards
Catalog No. 50-279-1818 Supplier Grainger Supplier No. 16D399
This wall-mount, freeze-resistant emergency shower is designed for climates where temperatures can drop below freezing. The unit features supply lines and a pre-built brass valve assembly that install in a heated space on the opposite side of a wall to prevent freezing, and the system drains completely after use to avoid standing water. It utilizes a hydrodynamic showerhead design that delivers a more effective and comfortable flow by distributing water equally across the entire footprint.
  • Flow rate of 20 GPM
  • Includes universal emergency sign and inspection tag
  • Galvanized steel pipe with 1 in. NPT female inlet
  • 11 in. diameter ABS plastic showerhead
  • Reduced installation time with pre-built valve assembly
  • Certified to ANSI Z358.1 and CSA standards