Learn More
Grainger Haws Axion MSR Emergency Drench Shower, Wall-Mounted, Corrosion-Resistant, 20 gpm, 1 ea
Catalog No. 502792257
Encompass_Preferred
The AXION® MSR corrosion-resistant emergency drench shower is designed for reliable performance in harsh environments. Featuring a heavy-duty, green epoxy-coated Schedule 80 PVC construction, this wall-mounted unit provides robust durability. It includes an integrated hydrodynamic showerhead that ensures uniform water distribution for effective decontamination. The system is certified to meet the ANSI/ISEA Z358.1 standard for emergency equipment.
  • Hydrodynamic ABS plastic showerhead delivers 20 gpm flow.
  • Corrosion-resistant construction with green epoxy coating.
  • Stainless steel type 316 ball valve with stainless steel ball and stem.
  • Includes 28 in. (71.1 cm) stainless steel triangular pull rod.
  • Designed for versatile horizontal or vertical installation.
  • Includes 8 in. x 10-3/4 in. (20.3 cm x 27.3 cm) universal emergency sign.
  • Pre-built valve assembly for reduced installati
Catalog No. 50-279-2257 Supplier Grainger Supplier No. 16D378
The AXION® MSR corrosion-resistant emergency drench shower is designed for reliable performance in harsh environments. Featuring a heavy-duty, green epoxy-coated Schedule 80 PVC construction, this wall-mounted unit provides robust durability. It includes an integrated hydrodynamic showerhead that ensures uniform water distribution for effective decontamination. The system is certified to meet the ANSI/ISEA Z358.1 standard for emergency equipment.
  • Hydrodynamic ABS plastic showerhead delivers 20 gpm flow.
  • Corrosion-resistant construction with green epoxy coating.
  • Stainless steel type 316 ball valve with stainless steel ball and stem.
  • Includes 28 in. (71.1 cm) stainless steel triangular pull rod.
  • Designed for versatile horizontal or vertical installation.
  • Includes 8 in. x 10-3/4 in. (20.3 cm x 27.3 cm) universal emergency sign.
  • Pre-built valve assembly for reduced installati