Learn More
Grainger Haws Emergency Drench Shower, Wall Mounted, Galvanized Steel, 1 in NPT, 1 ea
Catalog No. 502789872
Encompass_Preferred
This plumbed emergency drench shower provides an immediate response solution for environments where individuals may be exposed to injurious or corrosive materials. Designed for horizontal or vertical wall mounting, the unit features a high-performance drench showerhead with a hydrodynamic design that ensures equal water distribution. This model is certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergency eyewash and shower equipment.
  • Hydrodynamic ABS plastic drench showerhead with self-regulating 20 gpm flow control
  • Durable 1 in. Schedule 40 hot-dipped galvanized steel pipe construction
  • Chrome-plated brass stay-open ball valve with stainless steel ball and stem for corrosion resistance
  • 28 in. stainless steel triangular pull rod for reliable activation
  • Universal emergency sign and test card included for maintenance compliance
Catalog No. 50-278-9872 Supplier Grainger Supplier No. 16D376
This plumbed emergency drench shower provides an immediate response solution for environments where individuals may be exposed to injurious or corrosive materials. Designed for horizontal or vertical wall mounting, the unit features a high-performance drench showerhead with a hydrodynamic design that ensures equal water distribution. This model is certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergency eyewash and shower equipment.
  • Hydrodynamic ABS plastic drench showerhead with self-regulating 20 gpm flow control
  • Durable 1 in. Schedule 40 hot-dipped galvanized steel pipe construction
  • Chrome-plated brass stay-open ball valve with stainless steel ball and stem for corrosion resistance
  • 28 in. stainless steel triangular pull rod for reliable activation
  • Universal emergency sign and test card included for maintenance compliance