Learn More
Grainger Haws Emergency Drench Shower, Wall-Mounted, Stainless Steel, 20 gpm
Catalog No. 502792063
Encompass_Preferred
This corrosion-resistant emergency drench shower is designed for horizontal wall installation to provide immediate relief for personnel exposed to injurious or corrosive materials.
  • Features an AXION® MSR stainless steel showerhead with hydrodynamic flow distribution and a reliable stay-open ball valve mechanism for consistent operation.
  • Equipped with a Type 316 stainless steel stay-open ball valve for reliable flow activation
  • Includes a 28 in. (71.1 cm) stainless steel triangular pull rod
  • Constructed with 1 in. Schedule 40 stainless steel pipe for corrosion resistance
  • Certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergency eyewash and shower equipment
  • Includes an 8 in. x 10-3/4 in. (20.3 x 27.3 cm) universal emergency sign and maintenance test card
Catalog No. 50-279-2063 Supplier Grainger Supplier No. 16D402
This corrosion-resistant emergency drench shower is designed for horizontal wall installation to provide immediate relief for personnel exposed to injurious or corrosive materials.
  • Features an AXION® MSR stainless steel showerhead with hydrodynamic flow distribution and a reliable stay-open ball valve mechanism for consistent operation.
  • Equipped with a Type 316 stainless steel stay-open ball valve for reliable flow activation
  • Includes a 28 in. (71.1 cm) stainless steel triangular pull rod
  • Constructed with 1 in. Schedule 40 stainless steel pipe for corrosion resistance
  • Certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergency eyewash and shower equipment
  • Includes an 8 in. x 10-3/4 in. (20.3 x 27.3 cm) universal emergency sign and maintenance test card