Learn More
Northern Safety Emergency Shower Test Kit
SDP
Catalog No. NC1581300
Encompass
Vinyl sleeve fits over shower head and directs water into a bucket for easy clean-up. Includes sleeve with PVC handle for positioning, and heavy-duty 5-gallon bucket and lid for convenient storage. Comply with ANSI standards when testing emergency showers on a weekly schedule.
Catalog No. NC1581300 Supplier Northern Safety Supplier No. 6769
Vinyl sleeve fits over shower head and directs water into a bucket for easy clean-up. Includes sleeve with PVC handle for positioning, and heavy-duty 5-gallon bucket and lid for convenient storage. Comply with ANSI standards when testing emergency showers on a weekly schedule.