Learn More
GUARDIAN EQUIPMENT Test Kit for Emergency Showers and Safety Stations
Catalog No. NC0873131
Encompass_Preferred
Test emergency equipment for proper operation. Includes Tyvek chute and pail to contain tested water flow and prevent spillage.
Catalog No. NC0873131 Supplier GUARDIAN EQUIPMENT Supplier No. AP250005
Test emergency equipment for proper operation. Includes Tyvek chute and pail to contain tested water flow and prevent spillage.